Assay Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004897.1 | - | 13 - 14 | 1348 | 68 | |
| AK145900.1 | - | 13 - 14 | 1475 | 68 | ||
| AK149406.1 | - | 13 - 14 | 1353 | 68 | ||
| AK152772.1 | - | 13 - 14 | 1353 | 68 | ||
| AK153050.1 | - | 13 - 14 | 1481 | 68 | ||
| AK161189.1 | - | 13 - 14 | 1327 | 68 | ||
| AK168078.1 | - | 13 - 14 | 1353 | 68 | ||
| BC056644.1 | - | 13 - 14 | 1337 | 5098645 | 68 | |
| U15977.1 | - | 12 - 13 | 1266 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acsl1
Genomic Map