Assay Details
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113188.1 | NP_001106659.1 | 3 - 4 | 223 | 86 | |
| NM_001113189.1 | NP_001106660.1 | 3 - 4 | 223 | 86 | ||
| NM_008053.2 | NP_032079.1 | 3 - 4 | 223 | 86 | ||
| GenBank mRNA | BC019139.1 | - | 3 - 4 | 204 | 5041635 | 86 |
| CJ086949.1 | - | 3 - 4 | 224 | 86 | ||
| X90875.1 | - | 3 - 4 | 202 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fxr1
Genomic Map