Assay Details
| Gene Symbol: Ganab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293621.1 | NP_001280550.1 | 18 - 19 | 2305 | 98 | |
| NM_008060.2 | NP_032086.1 | 19 - 20 | 2371 | 98 | ||
| GenBank mRNA | AK017873.1 | - | 6 - 7 | 711 | 98 | |
| AK028655.1 | - | 18 - 19 | 2275 | 98 | ||
| AK030722.1 | - | 18 - 19 | 2274 | 98 | ||
| AK081915.1 | - | 18 - 19 | 2267 | 98 | ||
| AK122201.1 | - | 13 - 14 | 2501 | 98 | ||
| AK166150.1 | - | 18 - 19 | 2244 | 98 | ||
| BC020386.1 | - | 11 - 12 | 1521 | 98 | ||
| BC035423.1 | - | 9 - 10 | 1121 | 98 | ||
| BC094437.1 | - | 18 - 19 | 2255 | 30682580 | 98 | |
| BC117888.1 | - | 18 - 19 | 2248 | 40096413 | 98 | |
| BC117889.1 | - | 18 - 19 | 2251 | 40096414 | 98 | |
| BC128069.1 | - | 19 - 20 | 2315 | 40096428 | 98 | |
| U92793.1 | - | 19 - 20 | 2317 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ganab
Genomic Map