Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 4 - 5 | 1030 | 57 | |
| NM_008064.3 | NP_032090.3 | 4 - 5 | 1120 | 57 | ||
| GenBank mRNA | AK052211.1 | - | 4 - 5 | 1101 | 57 | |
| AK088481.1 | - | 4 - 5 | 1096 | 57 | ||
| AK139333.1 | - | 4 - 5 | 1068 | 57 | ||
| AK140042.1 | - | 4 - 5 | 1087 | 57 | ||
| AK141546.1 | - | 4 - 5 | 1103 | 57 | ||
| AK145631.1 | - | 3 - 4 | 535 | 57 | ||
| AK146538.1 | - | 4 - 5 | 1024 | 57 | ||
| AK150970.1 | - | 4 - 5 | 1078 | 57 | ||
| BC010210.1 | - | 4 - 5 | 1084 | 3969127 | 57 | |
| U49351.1 | - | 4 - 5 | 972 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map