Assay Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008078.2 | NP_032104.2 | 7 - 8 | 1176 | 58 | |
| GenBank mRNA | AF326549.1 | - | 7 - 8 | 840 | 58 | |
| AF326550.1 | - | 7 - 8 | 840 | 58 | ||
| AK136834.1 | - | 7 - 8 | 876 | 58 | ||
| BC018380.1 | - | 7 - 8 | 891 | 4482097 | 58 | |
| D42051.1 | - | 7 - 8 | 1192 | 58 | ||
| KC134211.1 | - | 7 - 8 | 840 | 58 | ||
| L16980.1 | - | 7 - 8 | 912 | 58 | ||
| S67454.1 | - | 3 - 4 | 318 | 58 | ||
More Information
Set Membership: |
|

Gad2
Genomic Map