Assay Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008078.2 | NP_032104.2 | 10 - 11 | 1431 | 86 | |
| GenBank mRNA | AF326549.1 | - | 10 - 11 | 1095 | 86 | |
| AF326550.1 | - | 10 - 11 | 1095 | 86 | ||
| AK136834.1 | - | 10 - 11 | 1131 | 86 | ||
| BC018380.1 | - | 10 - 11 | 1146 | 4482097 | 86 | |
| D42051.1 | - | 10 - 11 | 1447 | 86 | ||
| KC134211.1 | - | 10 - 11 | 1095 | 86 | ||
| L16980.1 | - | 10 - 11 | 1167 | 86 | ||
More Information
Set Membership: |
|

Gad2
Genomic Map