Assay Details
| Gene Symbol: Gba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077411.2 | NP_001070879.1 | 9 - 10 | 1505 | 59 | |
| NM_008094.5 | NP_032120.1 | 10 - 11 | 1674 | 59 | ||
| GenBank mRNA | AK082767.1 | - | 9 - 10 | 1503 | 59 | |
| AK152745.1 | - | 9 - 10 | 1486 | 59 | ||
| BC006663.1 | - | 10 - 11 | 1659 | 3498551 | 59 | |
| M24119.1 | - | 9 - 10 | 1414 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gba
Genomic Map