Assay Details
| Gene Symbol: Foxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008259.3 | NP_032285.2 | 1 - 2 | 455 | 68 | |
| GenBank mRNA | AB231452.1 | - | 1 - 2 | 74 | 68 | |
| BC096524.1 | - | 1 - 2 | 455 | 5720113 | 68 | |
| U44752.1 | - | 1 - 2 | 296 | 68 | ||
| X74936.1 | - | 1 - 2 | 262 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxa1
Genomic Map