Assay Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008304.2 | NP_032330.1 | 2 - 3 | 696 | 107 | |
| GenBank mRNA | AK011042.1 | - | 2 - 3 | 545 | 107 | |
| AK204318.1 | - | 1 - 3 | 107 | |||
| AK216657.1 | - | 1 - 2 | 123 | 107 | ||
| BC003929.1 | - | 1 - 2 | 94 | 107 | ||
| BC047144.1 | - | 2 - 3 | 562 | 6395969 | 107 | |
| U00674.1 | - | 2 - 3 | 696 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdc2
Genomic Map