Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 6 - 7 | 1021 | 68 | |
| GenBank mRNA | AK088493.1 | - | 6 - 7 | 1020 | 68 | |
| AK145520.1 | - | 6 - 7 | 1012 | 68 | ||
| AK146272.1 | - | 6 - 7 | 1018 | 68 | ||
| AK146770.1 | - | 6 - 7 | 1014 | 68 | ||
| AK146896.1 | - | 6 - 7 | 896 | 68 | ||
| AK150726.1 | - | 6 - 7 | 969 | 68 | ||
| AK210326.1 | - | 1 - 2 | 144 | 68 | ||
| BC050886.1 | - | 6 - 7 | 1029 | 5703922 | 68 | |
| BC051658.1 | - | 6 - 7 | 998 | 68 | ||
| BC057584.1 | - | 6 - 7 | 999 | 6401827 | 68 | |
| D86726.1 | - | 6 - 7 | 1021 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map