Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 7 - 8 | 1171 | 73 | |
| GenBank mRNA | AK078126.1 | - | 1 - 2 | 165 | 73 | |
| AK088493.1 | - | 7 - 8 | 1170 | 73 | ||
| AK145520.1 | - | 7 - 8 | 1162 | 73 | ||
| AK146272.1 | - | 7 - 8 | 1168 | 73 | ||
| AK146770.1 | - | 7 - 8 | 1164 | 73 | ||
| AK146896.1 | - | 7 - 8 | 1046 | 73 | ||
| AK150726.1 | - | 7 - 8 | 1119 | 73 | ||
| AK210326.1 | - | 2 - 3 | 294 | 73 | ||
| BC050886.1 | - | 7 - 8 | 1179 | 5703922 | 73 | |
| BC051658.1 | - | 7 - 8 | 1148 | 73 | ||
| BC057584.1 | - | 7 - 8 | 1149 | 6401827 | 73 | |
| D86726.1 | - | 7 - 8 | 1171 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map