Assay Details
| Gene Symbol: Men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168488.1 | NP_001161960.1 | 3 - 4 | 762 | 68 | |
| NM_001168489.1 | NP_001161961.1 | 3 - 4 | 740 | 68 | ||
| NM_001168490.1 | NP_001161962.1 | 2 - 3 | 1076 | 68 | ||
| NM_008583.2 | NP_032609.1 | 2 - 3 | 1076 | 68 | ||
| GenBank mRNA | AB023401.1 | - | 2 - 3 | 703 | 68 | |
| AF016398.1 | - | 3 - 4 | 715 | 68 | ||
| AF072755.1 | - | 3 - 4 | 699 | 68 | ||
| AF109389.1 | - | 2 - 3 | 1075 | 68 | ||
| AF130368.1 | - | 2 - 3 | 1058 | 68 | ||
| AK133984.1 | - | 3 - 4 | 699 | 68 | ||
| AK154371.1 | - | 3 - 4 | 721 | 68 | ||
| AK164700.1 | - | 3 - 4 | 697 | 68 | ||
| AK170713.1 | - | 2 - 3 | 1056 | 68 | ||
| BC006610.1 | - | 3 - 4 | 695 | 68 | ||
| BC036287.1 | - | 2 - 3 | 1004 | 4189611 | 68 | |
| CX568988.1 | - | 2 - 3 | 748 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Men1
Genomic Map