Assay Details
| Gene Symbol: Mep1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008586.2 | NP_032612.2 | 9 - 10 | 948 | 112 | |
| GenBank mRNA | AK098127.1 | - | 9 - 10 | 948 | 112 | |
| BC125627.1 | - | 9 - 10 | 946 | 40130142 | 112 | |
| BC145979.1 | - | 9 - 10 | 947 | 40131282 | 112 | |
| L15193.1 | - | 10 - 11 | 948 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mep1b
Genomic Map