Assay Details
| Gene Symbol: Mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008590.2 | NP_032616.1 | 1 - 2 | 287 | 55 | |
| GenBank mRNA | AK031718.1 | - | 1 - 2 | 286 | 55 | |
| AK032881.1 | - | 1 - 2 | 286 | 55 | ||
| AK034949.1 | - | 1 - 2 | 285 | 55 | ||
| AK168714.1 | - | 1 - 2 | 285 | 55 | ||
| AK168732.1 | - | 1 - 2 | 285 | 55 | ||
| AK168743.1 | - | 1 - 2 | 285 | 55 | ||
| AK168995.1 | - | 1 - 2 | 285 | 55 | ||
| AK169095.1 | - | 1 - 2 | 285 | 55 | ||
| AK169266.1 | - | 1 - 2 | 285 | 55 | ||
| BC006639.1 | - | 1 - 2 | 270 | 3482928 | 55 | |
| CJ162886.1 | - | 1 - 2 | 288 | 55 | ||
| D16262.1 | - | 1 - 2 | 274 | 55 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032881.1,AK034949.1,AK168714.1,AK168732.1,AK168743.1,AK168995.1,AK169095.1,AK169266.1,BC006639.1,D16262.1,NM_008590.2 transcripts.
Set Membership: |
|

Mest
Genomic Map