Assay Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008618.3 | NP_032644.3 | 7 - 8 | 994 | 74 | |
| GenBank mRNA | AK002404.1 | - | 7 - 8 | 852 | 74 | |
| AK005237.1 | - | 7 - 8 | 889 | 74 | ||
| AK008270.1 | - | 7 - 8 | 866 | 74 | ||
| AK164785.1 | - | 7 - 8 | 875 | 74 | ||
| AK168545.1 | - | 7 - 8 | 832 | 74 | ||
| BC050940.1 | - | 7 - 8 | 860 | 6820601 | 74 | |
| M29462.1 | - | 7 - 8 | 873 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdh1
Genomic Map