Assay Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008623.4 | NP_032649.2 | 5 - 6 | 621 | 68 | |
| GenBank mRNA | AK089180.1 | - | 5 - 6 | 625 | 68 | |
| AK157960.1 | - | 3 - 4 | 518 | 68 | ||
| BC139139.1 | - | 3 - 4 | 486 | 8862161 | 68 | |
| BC141226.1 | - | 3 - 4 | 486 | 9055838 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpz
Genomic Map