Assay Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008625.2 | NP_032651.2 | 8 - 9 | 1499 | 74 | |
| GenBank mRNA | BC141338.1 | - | 8 - 9 | 1451 | 9055950 | 74 |
| Z11974.1 | - | 8 - 9 | 1499 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrc1
Genomic Map