Assay Details
| Gene Symbol: Mut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008650.3 | NP_032676.2 | 6 - 7 | 1459 | 73 | |
| GenBank mRNA | AK146309.1 | - | 6 - 7 | 1419 | 73 | |
| X51941.1 | - | 5 - 6 | 1370 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mut
Genomic Map