Assay Details
| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093775.1 | NP_001087244.1 | 17 - 18 | 3557 | 86 | |
| NM_001093776.1 | NP_001087245.1 | 17 - 18 | 3463 | 86 | ||
| NM_001093778.1 | NP_001087247.1 | 17 - 18 | 3548 | 86 | ||
| NM_008666.3 | NP_032692.2 | 17 - 18 | 3551 | 86 | ||
| GenBank mRNA | AB093283.1 | - | 14 - 15 | 2910 | 86 | |
| AB212898.1 | - | 16 - 17 | 3155 | 86 | ||
| AF004295.1 | - | 17 - 18 | 3367 | 86 | ||
| AK049967.1 | - | 8 - 9 | 1392 | 86 | ||
| AK076608.1 | - | 5 - 6 | 988 | 86 | ||
| AK161925.1 | - | 1 - 2 | 92 | 86 | ||
| BC079651.1 | - | 17 - 18 | 3432 | 86 | ||
| BC094438.1 | - | 13 - 14 | 2788 | 30692780 | 86 | |
| BC131677.1 | - | 16 - 17 | 3152 | 40111036 | 86 | |
| BQ876229.1 | - | 4 - 5 | 684 | 85 | ||
| U86338.1 | - | 17 - 18 | 3279 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myt1l
Genomic Map