Assay Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009009.4 | NP_033035.3 | 3 - 4 | 517 | 87 | |
| GenBank mRNA | AF332085.1 | - | 2 - 3 | 272 | 87 | |
| AF332086.1 | - | 2 - 3 | 272 | 87 | ||
| AK004746.1 | - | 3 - 4 | 320 | 87 | ||
| AK128984.1 | - | 3 - 4 | 406 | 87 | ||
| AK139492.1 | - | 2 - 3 | 170 | 87 | ||
| AK145591.1 | - | 3 - 4 | 484 | 87 | ||
| AK163992.1 | - | 378 | 87 | |||
| BC043032.1 | - | 2 - 3 | 286 | 5696540 | 87 | |
| BC129916.1 | - | 2 - 3 | 316 | 40111820 | 87 | |
| BC129917.1 | - | 2 - 3 | 317 | 40111821 | 87 | |
| X98293.1 | - | 2 - 3 | 272 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad21
Genomic Map