Assay Details
| Gene Symbol: Rad54l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122958.1 | NP_001116430.1 | 2 - 3 | 292 | 59 | |
| NM_001122959.1 | NP_001116431.1 | 2 - 3 | 271 | 59 | ||
| NM_009015.3 | NP_033041.3 | 1 - 2 | 622 | 59 | ||
| GenBank mRNA | AK017045.1 | - | 2 - 3 | 294 | 59 | |
| AK030753.1 | - | 2 - 3 | 250 | 59 | ||
| AK088876.1 | - | 2 - 3 | 289 | 59 | ||
| AK161644.1 | - | 2 - 3 | 245 | 59 | ||
| AK185368.1 | - | 1 - 2 | 235 | 59 | ||
| AK186969.1 | - | 1 - 2 | 236 | 59 | ||
| BY716729.1 | - | 2 - 3 | 292 | 59 | ||
| CN669448.1 | - | 1 - 2 | 580 | 59 | ||
| X97796.1 | - | 1 - 2 | 82 | 59 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK088876.1,NM_001122958.1,NM_001122959.1,NM_009015.3,X97796.1 transcripts.
Set Membership: |
|

Rad54l
Genomic Map