Assay Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009029.2 | NP_033055.2 | 7 - 8 | 842 | 82 | |
| GenBank mRNA | AK011246.1 | - | 7 - 8 | 842 | 82 | |
| AK141028.1 | - | 7 - 8 | 874 | 82 | ||
| AK146467.1 | - | 2 - 3 | 173 | 82 | ||
| BC096525.1 | - | 7 - 8 | 842 | 6832808 | 82 | |
| DQ400415.1 | - | 7 - 8 | 692 | 82 | ||
| M26391.1 | - | 7 - 8 | 790 | 82 | ||
More Information
Set Membership: |
|

Rb1
Genomic Map