Assay Details
| Gene Symbol: Rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009044.2 | NP_033070.2 | 1 - 2 | 296 | 99 | |
| GenBank mRNA | AK031627.1 | - | 1 - 2 | 95 | 99 | |
| BC139447.1 | - | 1 - 2 | 236 | 8862469 | 99 | |
| BC139448.1 | - | 1 - 2 | 236 | 8862470 | 99 | |
| BC139770.1 | - | 1 - 2 | 238 | 40111461 | 99 | |
| X15842.1 | - | 1 - 2 | 415 | 99 | ||
| X60271.1 | - | 1 - 2 | 295 | 99 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC139447.1,BC139448.1,BC139770.1,NM_009044.2,X15842.1,X60271.1 transcripts.
Set Membership: |
|

Rel
Genomic Map