Assay Details
| Gene Symbol: Relb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290457.1 | NP_001277386.1 | 3 - 4 | 628 | 103 | |
| NM_009046.2 | NP_033072.2 | 4 - 5 | 637 | 103 | ||
| GenBank mRNA | AK146932.1 | - | 4 - 5 | 638 | 103 | |
| AK156767.1 | - | 4 - 5 | 593 | 103 | ||
| BC019765.1 | - | 4 - 5 | 637 | 3584200 | 103 | |
| BC034523.1 | - | 3 - 4 | 441 | 4485540 | 103 | |
| BC117792.2 | - | 8 - 9 | 103 | |||
| BC117793.1 | - | 4 - 5 | 573 | 40092162 | 103 | |
| DQ020179.1 | - | 3 - 4 | 438 | 103 | ||
| DQ020181.1 | - | 4 - 5 | 447 | 103 | ||
| M83380.1 | - | 4 - 5 | 474 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Relb
Genomic Map