Assay Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009101.2 | NP_033127.1 | 3 - 4 | 349 | 70 | |
| GenBank mRNA | AK155855.1 | - | 3 - 4 | 379 | 70 | |
| BC009105.1 | - | 3 - 4 | 366 | 3483313 | 70 | |
| BC110562.1 | - | 3 - 4 | 351 | 40049793 | 70 | |
| BC110563.1 | - | 3 - 4 | 351 | 40049794 | 70 | |
| BC145932.1 | - | 3 - 4 | 395 | 40131235 | 70 | |
| BC145934.1 | - | 3 - 4 | 395 | 40131237 | 70 | |
| M21019.1 | - | 3 - 4 | 351 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rras
Genomic Map