Assay Details
| Gene Symbol: Rrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009103.3 | NP_033129.2 | 17 - 18 | 2331 | 92 | |
| GenBank mRNA | AK082135.1 | - | 17 - 18 | 2209 | 92 | |
| AK088043.1 | - | 17 - 18 | 2178 | 92 | ||
| AK137075.1 | - | 17 - 18 | 2164 | 92 | ||
| AK168586.1 | - | 17 - 18 | 2198 | 92 | ||
| AK200772.1 | - | 1 - 2 | 60 | 92 | ||
| AK203165.1 | - | 1 - 2 | 60 | 92 | ||
| AK209968.1 | - | 1 - 2 | 60 | 92 | ||
| BC003299.1 | - | 17 - 18 | 2199 | 92 | ||
| BC016450.1 | - | 17 - 18 | 2159 | 4485739 | 92 | |
| BC066217.1 | - | 17 - 18 | 2209 | 30055026 | 92 | |
| K02927.1 | - | 17 - 18 | 2082 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rrm1
Genomic Map