Assay Details
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009118.2 | NP_033144.1 | 1 - 2 | 336 | 63 | |
| GenBank mRNA | AK053278.1 | - | 1 - 2 | 95 | 63 | |
| AK143098.1 | - | 1 - 2 | 95 | 63 | ||
| AK149299.1 | - | 1 - 2 | 345 | 63 | ||
| AY651760.1 | - | 1 - 2 | 124 | 63 | ||
| BC016498.1 | - | 1 - 2 | 129 | 4527551 | 63 | |
| M24086.1 | - | 1 - 2 | 123 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK053278.1,AK143098.1,AK149299.1,AY651760.1,BC016498.1,M24086.1,NM_009118.2 transcripts.
Set Membership: |
|

Sag
Genomic Map