Assay Details
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009118.2 | NP_033144.1 | 8 - 9 | 1008 | 85 | |
| GenBank mRNA | AK044214.1 | - | 7 - 8 | 689 | 85 | |
| AK053278.1 | - | 8 - 9 | 767 | 85 | ||
| AK143098.1 | - | 8 - 9 | 767 | 85 | ||
| AK149299.1 | - | 8 - 9 | 1017 | 85 | ||
| AY651760.1 | - | 8 - 9 | 796 | 85 | ||
| BC016498.1 | - | 8 - 9 | 801 | 4527551 | 85 | |
| M24086.1 | - | 8 - 9 | 795 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sag
Genomic Map