Assay Details
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009118.2 | NP_033144.1 | 11 - 12 | 1303 | 69 | |
| GenBank mRNA | AK044214.1 | - | 10 - 11 | 984 | 69 | |
| AK143098.1 | - | 11 - 12 | 1062 | 69 | ||
| AK149299.1 | - | 11 - 12 | 1312 | 69 | ||
| BC016498.1 | - | 11 - 12 | 1096 | 4527551 | 69 | |
| BC026984.1 | - | 2 - 3 | 2114 | 69 | ||
| M24086.1 | - | 11 - 12 | 1090 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sag
Genomic Map