Assay Details
| Gene Symbol: Shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009168.2 | NP_033194.1 | 2 - 3 | 563 | 98 | |
| GenBank mRNA | AB018423.1 | - | 2 - 3 | 562 | 98 | |
| AK142491.1 | - | 2 - 3 | 579 | 98 | ||
| AK164639.1 | - | 2 - 3 | 499 | 98 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253054 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62458 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253038 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03338723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08810020 | - | - |
| Whole Mouse Genome G4122A | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | AB018423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | BC079574.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | BC103798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | AK142491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374818 | AK164639.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110724 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13296353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13188617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13296362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13343677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13188616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253054 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13068795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13119433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13343678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13068798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13296354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13068802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13061981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13296356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13188613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445998 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417486_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445992 | - | - |
| Mouse Genome 430 2.0 Array | 1417486_at | - | - |
| Mouse Genome 430 2.0 Array | 1417486_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20420_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20420_at | - | - |
| Mouse Expression Set 430 | 1417486_PM_at | - | - |
| Mouse Expression Set 430 | 1417486_at | - | - |
| Murine Genome U74v2 Set | 103819_at | - | - |
| Murine Genome U74v2 Set | 103819_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417486_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417486_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417486_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417486_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 560089 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3390193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3390193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775742 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775742 | - | - |
| mouse-6 v1.1 (Illumina) | scl50606.7.1_142-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390193 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50606.7.1_142-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2858666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775742 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2858666 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50606.7.1_142-S | - | - |

Shd
Genomic Map