Assay Details
| Gene Symbol: Shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159523.1 | NP_001152995.1 | 3 - 4 | 680 | 96 | |
| NM_009168.2 | NP_033194.1 | 3 - 4 | 677 | 96 | ||
| GenBank mRNA | AB018423.1 | - | 3 - 4 | 676 | 96 | |
| AK142491.1 | - | 3 - 4 | 693 | 96 | ||
| AK164639.1 | - | 3 - 4 | 613 | 96 | ||
| BC103798.1 | - | 3 - 4 | 573 | 30288676 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shd
Genomic Map