| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600014417.mm.1 |
TC0600001821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600014417.mm.1 |
NM_009169.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014417.mm.1 |
NM_009169.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014420.mm.1 |
TC0600001821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600014420.mm.1 |
NM_009169.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014420.mm.1 |
NM_009169.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014418.mm.1 |
TC0600001821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600014418.mm.1 |
NM_009169.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014418.mm.1 |
NM_009169.2 |
Refseq |