Assay Details
| Gene Symbol: Siah1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CJ063158.1 | - | 1 - 2 | 111 | 63 | |
| Z19580.1 | - | 1 - 2 | 39 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the Z19580.1 transcripts.
This assay is in the 5'UTR region of the Z19580.1 transcripts.
Set Membership: |
|

Siah1b
Genomic Map