Assay Details
| Gene Symbol: Siah1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC052887.1 | - | 2 - 3 | 128 | 30053109 | 103 |
| CF723667.1 | - | 2 - 3 | 384 | 103 | ||
| CJ063158.1 | - | 2 - 3 | 205 | 103 | ||
| Z19580.1 | - | 2 - 3 | 133 | 103 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC052887.1,Z19580.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796349 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796350 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796348 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796347 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796354 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796352 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972182 | Z19580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972182 | NM_009173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972182 | BC052887.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138399 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152668 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972182 | Z19580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972182 | NM_009173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972182 | BC052887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P44994 | Z19580.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P44994 | NM_009173.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P44994 | BC052887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378602 | Z19580.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378602 | Z19579.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378602 | NM_009173.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378602 | BC052887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378602 | NM_009172.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32796380 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152668 | - | - |
| Whole Mouse Genome G4122A | A_52_P44994 | Z19580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P44994 | NM_009173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P44994 | BC052887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378602 | Z19580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378602 | Z19579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378602 | NM_009173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378602 | BC052887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378602 | NM_009172.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113377 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918310 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796348 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796352 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796347 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796349 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796354 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32796350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32796286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14617650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14518297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14625182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14680828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32796362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607748 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607750 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607747 | - | - |
| Mouse Genome 430 2.0 Array | 1419404_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1419404_PM_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00020740.mm.1 | NM_009173.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020740.mm.1 | TC0X00003329.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020741.mm.1 | NM_009173.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020741.mm.1 | TC0X00003329.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419404_PM_s_at | - | - |
| Mouse Expression Set 430 | 1419404_s_at | - | - |
| Murine Genome U74v2 Set | 92290_at | - | - |
| Murine Genome U74v2 Set | 92290_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419404_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419404_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 750302 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200520 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6290519 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1170309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217391 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646574 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706393 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217391 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2646574 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618014 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706393 | - | - |
| mouse-6 v1.1 (Illumina) | scl020438.3_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl53613.6.1_106-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0020438.2_18-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0020438.2_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050279 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53613.6.1_106-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020438.2_182-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020438.3_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020438.2_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290519 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200520 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050279 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2706393 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1217391 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2646574 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2903734 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706393 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217391 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2646574 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2903734 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618014 | - | - |

Siah1b
Genomic Map