Assay Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009180.3 | NP_033206.2 | 3 - 4 | 458 | 88 | |
| GenBank mRNA | AK004613.1 | - | 3 - 4 | 459 | 88 | |
| AK136381.1 | - | 461 | 88 | |||
| BC003276.1 | - | 3 - 4 | 1360 | 88 | ||
| BC010208.1 | - | 410 | 88 | |||
| X93999.1 | - | 3 - 4 | 422 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St6galnac2
Genomic Map