Assay Details
| Gene Symbol: Sox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009238.2 | NP_033264.2 | 1 | 1556 | 82 | |
| GenBank mRNA | AK028989.1 | - | 1572 | 82 | ||
| BC052736.1 | - | 1 | 1556 | 6822618 | 82 | |
| BC060669.1 | - | 1 | 1547 | 82 | ||
| X70298.1 | - | 1 | 1551 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox4
Genomic Map