Assay Details
| Gene Symbol: Sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK159249.1 | - | 6 - 7 | 583 | 68 | |
| BC004638.1 | - | 6 - 7 | 528 | 3586402 | 68 | |
| CT010306.1 | - | 5 - 6 | 450 | 68 | ||
| X04017.1 | - | 6 - 7 | 539 | 68 | ||
| X12697.1 | - | 1 - 2 | 87 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sparc
Genomic Map