Assay Details
| Gene Symbol: Sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290817.1 | NP_001277746.1 | 8 - 9 | 1032 | 103 | |
| NM_009242.5 | NP_033268.1 | 8 - 9 | 1035 | 103 | ||
| GenBank mRNA | AK014286.1 | - | 8 - 9 | 835 | 103 | |
| AK049692.1 | - | 8 - 9 | 821 | 103 | ||
| AK082283.1 | - | 7 - 8 | 870 | 103 | ||
| AK133377.1 | - | 8 - 9 | 805 | 103 | ||
| AK146935.1 | - | 8 - 9 | 805 | 103 | ||
| AK148670.1 | - | 8 - 9 | 820 | 103 | ||
| AK161229.1 | - | 8 - 9 | 823 | 103 | ||
| BC004638.1 | - | 8 - 9 | 807 | 3586402 | 103 | |
| CT010306.1 | - | 7 - 8 | 729 | 103 | ||
| X04017.1 | - | 8 - 9 | 818 | 103 | ||
| X12697.1 | - | 3 - 4 | 365 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sparc
Genomic Map