Assay Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009261.3 | NP_033287.2 | 5 - 6 | 794 | 78 | |
| GenBank mRNA | BC014710.1 | - | 6 - 7 | 878 | 4502840 | 78 |
| X84692.1 | - | 731 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Strbp
Genomic Map