Assay Details
| Gene Symbol: Srpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009274.2 | NP_033300.2 | 8 - 9 | 980 | 102 | |
| GenBank mRNA | AB006036.1 | - | 8 - 9 | 773 | 102 | |
| AK136861.1 | - | 8 - 9 | 946 | 102 | ||
| AK196829.1 | - | 2 - 3 | 96 | 102 | ||
| BC020178.1 | - | 8 - 9 | 877 | 4507346 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk2
Genomic Map