Assay Details
| Gene Symbol: Srpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009274.2 | NP_033300.2 | 9 - 10 | 1233 | 87 | |
| GenBank mRNA | AB006036.1 | - | 9 - 10 | 1023 | 87 | |
| AK136861.1 | - | 9 - 10 | 1199 | 87 | ||
| BC020178.1 | - | 9 - 10 | 1130 | 4507346 | 87 | |
| BC062941.1 | - | 9 - 10 | 1152 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srpk2
Genomic Map