Assay Details
| Gene Symbol: Supt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009297.2 | NP_033323.2 | 6 - 7 | 811 | 107 | |
| GenBank mRNA | AK136094.1 | - | 1451 | 107 | ||
| AK170533.1 | - | 6 - 7 | 810 | 107 | ||
| BC072657.1 | - | 6 - 7 | 731 | 30536338 | 107 | |
| U40375.1 | - | 6 - 7 | 809 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt6
Genomic Map