Assay Details
| Gene Symbol: Svs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009301.2 | NP_033327.1 | 2 - 3 | 431 | 62 | |
| GenBank mRNA | AK007282.1 | - | 2 - 3 | 432 | 62 | |
| BC114349.1 | - | 2 - 3 | 435 | 40053311 | 62 | |
| S60687.1 | - | 1 - 2 | 321 | 62 | ||
| X57139.1 | - | 2 - 3 | 459 | 62 | ||
| X63161.1 | - | 2 - 3 | 411 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | X57139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | X63161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | X57139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353162 | X63161.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881821 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107085 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | X57139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | X63161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | X57139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353162 | X63161.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21817303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107085 | - | - |
| Whole Mouse Genome G4122A | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | X57139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | X63161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | NM_009301.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | AK007282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | X57139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353162 | X63161.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130223 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01598887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21817331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01598878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01598902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02248829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01315414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01598887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21817278 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478471 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478470 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419347_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427220_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438616_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456124_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419346_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478469 | - | - |
| Mouse Genome 430 2.0 Array | 1419347_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427220_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419346_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419347_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438616_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1456124_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419346_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427220_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456124_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438616_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20944_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20944_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200020486.mm.1 | NM_009301.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020486.mm.1 | NM_009301.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020486.mm.1 | TC0200002513.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200020487.mm.1 | NM_009301.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020487.mm.1 | NM_009301.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200020487.mm.1 | TC0200002513.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419346_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438616_x_at | - | - |
| Mouse Expression Set 430 | 1456124_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427220_a_at | - | - |
| Mouse Expression Set 430 | 1419347_x_at | - | - |
| Mouse Expression Set 430 | 1419347_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438616_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427220_PM_a_at | - | - |
| Mouse Expression Set 430 | 1419346_a_at | - | - |
| Mouse Expression Set 430 | 1456124_x_at | - | - |
| Murine Genome U74v2 Set | 98842_PM_at | - | - |
| Murine Genome U74v2 Set | 98842_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419347_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427220_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438616_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419347_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456124_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419346_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456124_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438616_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419346_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427220_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419347_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427220_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419347_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438616_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456124_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419346_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419346_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438616_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427220_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456124_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 515131 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4280041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227330 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227330 | - | - |
| mouse-6 v1.1 (Illumina) | scl19941.3.19_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19941.3.19_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227330 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227330 | - | - |

Svs5
Genomic Map