Assay Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 5 - 6 | 856 | 92 | |
| GenBank mRNA | AF015038.1 | - | 5 - 6 | 802 | 92 | |
| AK052141.1 | - | 5 - 6 | 831 | 92 | ||
| AK053832.1 | - | 5 - 6 | 2142 | 92 | ||
| AK150695.1 | - | 5 - 6 | 830 | 92 | ||
| AK152094.1 | - | 5 - 6 | 830 | 92 | ||
| AK152790.1 | - | 5 - 6 | 831 | 92 | ||
| AK152994.1 | - | 5 - 6 | 831 | 92 | ||
| AK154855.1 | - | 5 - 6 | 870 | 92 | ||
| BC054726.1 | - | 5 - 6 | 856 | 5703563 | 92 | |
| D38117.1 | - | 5 - 6 | 897 | 92 | ||
| Y10139.1 | - | 5 - 6 | 738 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn2
Genomic Map