Assay Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 8 - 9 | 1104 | 93 | |
| GenBank mRNA | AF015038.1 | - | 8 - 9 | 1050 | 93 | |
| AK052141.1 | - | 8 - 9 | 1079 | 93 | ||
| AK053832.1 | - | 8 - 9 | 2390 | 93 | ||
| AK078787.1 | - | 4 - 5 | 1733 | 93 | ||
| AK150695.1 | - | 8 - 9 | 1078 | 93 | ||
| AK152094.1 | - | 8 - 9 | 1078 | 93 | ||
| AK152790.1 | - | 8 - 9 | 1080 | 93 | ||
| AK152994.1 | - | 8 - 9 | 1080 | 93 | ||
| AK154855.1 | - | 8 - 9 | 1118 | 93 | ||
| BC054726.1 | - | 8 - 9 | 1104 | 5703563 | 93 | |
| D38117.1 | - | 8 - 9 | 1145 | 93 | ||
| Y10139.1 | - | 8 - 9 | 986 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn2
Genomic Map