Assay Details
| Gene Symbol: Capzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037761.2 | NP_001032850.1 | 6 - 7 | 708 | 70 | |
| NM_001271405.1 | NP_001258334.1 | 6 - 7 | 687 | 70 | ||
| NM_001271406.1 | NP_001258335.1 | 6 - 7 | 673 | 70 | ||
| NM_009798.4 | NP_033928.1 | 6 - 7 | 708 | 70 | ||
| GenBank mRNA | AK153367.1 | - | 6 - 7 | 485 | 70 | |
| AK153394.1 | - | 6 - 7 | 485 | 70 | ||
| AK156778.1 | - | 6 - 7 | 677 | 70 | ||
| AK159390.1 | - | 6 - 7 | 682 | 70 | ||
| AK160082.1 | - | 6 - 7 | 683 | 70 | ||
| AK162757.1 | - | 6 - 7 | 675 | 70 | ||
| AK168738.1 | - | 6 - 7 | 636 | 70 | ||
| AK177260.1 | - | 1 - 2 | 36 | 70 | ||
| AK182951.1 | - | 1 - 2 | 36 | 70 | ||
| AK186717.1 | - | 1 - 2 | 36 | 70 | ||
| AK187793.1 | - | 1 - 2 | 36 | 70 | ||
| AK188337.1 | - | 1 - 2 | 36 | 70 | ||
| AK192076.1 | - | 1 - 2 | 36 | 70 | ||
| AK198394.1 | - | 2 - 3 | 126 | 70 | ||
| AK200176.1 | - | 1 - 2 | 36 | 70 | ||
| AK207588.1 | - | 1 - 2 | 36 | 70 | ||
| AK212957.1 | - | 1 - 2 | 99 | 70 | ||
| BC002053.1 | - | 6 - 7 | 650 | 3590980 | 70 | |
| FJ692320.1 | - | 6 - 7 | 673 | 70 | ||
| U10406.1 | - | 6 - 7 | 628 | 70 | ||
| U10407.1 | - | 6 - 7 | 599 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capzb
Genomic Map