Assay Details
| Gene Symbol: Car1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083957.1 | NP_001077426.1 | 1 - 2 | 109 | 72 | |
| NM_009799.4 | NP_033929.2 | 2 - 3 | 121 | 72 | ||
| GenBank mRNA | AK003066.1 | - | 2 - 3 | 122 | 72 | |
| AK033719.1 | - | 1 - 2 | 110 | 72 | ||
| AK146372.1 | - | 2 - 3 | 122 | 72 | ||
| AK162331.1 | - | 1 - 2 | 109 | 72 | ||
| AK172076.1 | - | 2 - 3 | 122 | 72 | ||
| BB842334.1 | - | 2 - 3 | 122 | 72 | ||
| BC011223.1 | - | 1 - 2 | 93 | 4208180 | 72 | |
| BC110681.1 | - | 1 - 2 | 91 | 4984216 | 72 | |
| BC132432.1 | - | 2 - 3 | 77 | 40130709 | 72 | |
| BC132434.1 | - | 2 - 3 | 77 | 40130711 | 72 | |
| M32452.1 | - | 1 - 2 | 78 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003066.1, AK146372.1, AK162331.1, AK172076.1, BC011223.1, BC110681.1, BC132432.1, BC132434.1, M32452.1, NM_001083957.1, NM_009799.4 transcripts.
Set Membership: |
|

Car1
Genomic Map