Assay Details
| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009818.1 | NP_033948.1 | 8 - 9 | 1216 | 67 | |
| GenBank mRNA | AK004623.1 | - | 8 - 9 | 1174 | 67 | |
| AK036752.1 | - | 8 - 9 | 1241 | 67 | ||
| AK046513.1 | - | 2 - 3 | 575 | 67 | ||
| AK148496.1 | - | 8 - 9 | 1213 | 67 | ||
| AK149721.1 | - | 8 - 9 | 1231 | 67 | ||
| AK149738.1 | - | 8 - 9 | 1231 | 67 | ||
| AK159339.1 | - | 8 - 9 | 1231 | 67 | ||
| AK168635.1 | - | 8 - 9 | 1241 | 67 | ||
| BC048163.1 | - | 8 - 9 | 1204 | 6418358 | 67 | |
| BC060174.1 | - | 8 - 9 | 1211 | 67 | ||
| BC068316.1 | - | 8 - 9 | 1208 | 30106659 | 67 | |
| BC105576.1 | - | 8 - 9 | 1196 | 3967124 | 67 | |
| D90362.1 | - | 8 - 9 | 1207 | 67 | ||
| X59990.1 | - | 8 - 9 | 1216 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnna1
Genomic Map