Assay Details
| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009818.1 | NP_033948.1 | 9 - 10 | 1368 | 125 | |
| GenBank mRNA | AK004623.1 | - | 9 - 10 | 1326 | 125 | |
| AK036752.1 | - | 9 - 10 | 1393 | 125 | ||
| AK046513.1 | - | 3 - 4 | 727 | 125 | ||
| AK148496.1 | - | 9 - 10 | 1365 | 125 | ||
| AK149721.1 | - | 9 - 10 | 1383 | 125 | ||
| AK149738.1 | - | 9 - 10 | 1383 | 125 | ||
| AK159339.1 | - | 9 - 10 | 1383 | 125 | ||
| AK168635.1 | - | 9 - 10 | 1393 | 125 | ||
| BC048163.1 | - | 9 - 10 | 1356 | 6418358 | 125 | |
| BC060174.1 | - | 9 - 10 | 1363 | 125 | ||
| BC068316.1 | - | 9 - 10 | 1360 | 30106659 | 125 | |
| BC105576.1 | - | 9 - 10 | 1348 | 3967124 | 125 | |
| D90362.1 | - | 9 - 10 | 1359 | 125 | ||
| X59990.1 | - | 9 - 10 | 1368 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnna1
Genomic Map