Assay Details
| Gene Symbol: Runx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111022.2 | NP_001104492.1 | 5 - 6 | 774 | 110 | |
| NM_009821.3 | NP_033951.2 | 3 - 4 | 2109 | 110 | ||
| GenBank mRNA | AK051758.1 | - | 3 - 4 | 1647 | 110 | |
| BC069929.1 | - | 5 - 6 | 711 | 4008335 | 110 | |
| D26532.1 | - | 3 - 4 | 2106 | 110 | ||
More Information
Set Membership: |
|

Runx1
Genomic Map